Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134247.3 | NP_599008.3 | 3 | 2204 | ||
GenBank mRNA | AK046630.1 | - | 3 | 2204 |