Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161355.1 | NP_001154827.1 | 9 | 1178 | ||
NM_001161356.1 | NP_001154828.1 | 9 | 951 | |||
NM_134249.5 | NP_599010.4 | 9 | 1060 | |||
GenBank mRNA | AB009015.1 | - | 9 | 951 | ||
AF399827.1 | - | 9 | 912 | |||
AF399828.1 | - | 9 | 912 | |||
AK145630.1 | - | 9 | 939 | |||
AK163225.1 | - | 9 | 939 | |||
AK166877.1 | - | 9 | 932 | |||
BC025096.1 | - | 9 | 990 | 4158605 | ||
BC028829.1 | - | 9 | 1022 | 5102129 |