Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134255.3 | NP_599016.2 | 8 | 1968 | ||
GenBank mRNA | AK004319.1 | - | 1 | 563 | ||
AK028761.1 | - | 8 | 1907 | |||
AK031743.1 | - | 8 | 1971 | |||
AK045274.1 | - | 8 | 1947 | |||
AK051580.1 | - | 8 | 1966 | |||
AK085663.1 | - | 8 | 1931 | |||
AK085696.1 | - | 8 | 1918 | |||
AK088153.1 | - | 8 | 1907 | |||
AK162451.1 | - | 8 | 1969 | |||
AK167755.1 | - | 8 | 1909 | |||
AK168360.1 | - | 8 | 1909 | |||
AK168987.1 | - | 8 | 1896 | |||
BC022911.1 | - | 8 | 1877 | 4457285 |