Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134471.4 | NP_608301.3 | 2, 3 | 289 | ||
GenBank mRNA | AK015046.1 | - | 2, 3 | 259 | ||
AK088047.1 | - | 2, 3 | 230 | |||
AK150222.1 | - | 2, 3 | 220 | |||
BC006841.1 | - | 2, 3 | 249 | 3598157 | ||
CF163926.1 | - | 2, 3 | 289 |