Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199349.1 | NP_001186278.1 | 10 | 3371 | ||
NM_138309.3 | NP_612182.1 | 9 | 3302 | |||
GenBank mRNA | AK009181.1 | - | 1 | 1505 | ||
AK080225.1 | - | 10 | 3127 | |||
AK166275.1 | - | 10 | 3114 | |||
AK189156.1 | - | 1 | 209 | |||
AK206730.1 | - | 1 | 209 | |||
AY078163.1 | - | 9 | 3096 | |||
BC029837.1 | - | 1 | 1110 |