Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162423.1 | NP_001155895.1 | 21 | 2664 | ||
NM_001162424.1 | NP_001155896.1 | 20 | 2634 | |||
NM_138630.2 | NP_619536.2 | 21 | 2697 | |||
GenBank mRNA | AF443826.1 | - | 21 | 2628 | ||
AK040687.1 | - | 14 | 1503 | |||
AK129060.1 | - | 3206 | ||||
AK157294.1 | - | 21 | 2696 | |||
AK169865.1 | - | 18 | 2132 | |||
AY183124.1 | - | 21 | 2670 | |||
AY183125.1 | - | 20 | 2607 | |||
AY183126.1 | - | 20 | 2764 | |||
AY183127.1 | - | 21 | 2670 | |||
AY183128.1 | - | 17 | 2359 | |||
AY183129.1 | - | 21 | 2679 | |||
AY183130.1 | - | 17 | 2042 | |||
AY183131.1 | - | 16 | 2136 | |||
AY183132.1 | - | 15 | 2362 | |||
AY183133.1 | - | 19 | 2703 | |||
BC109001.1 | - | 21 | 2595 | 40048081 | ||
BC109002.1 | - | 19 | 2654 |