Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138650.2 | NP_619591.1 | 3 | 591 | ||
GenBank mRNA | AK083258.1 | - | 3 | 584 | ||
AK087972.1 | - | 3 | 549 | |||
AK089261.1 | - | 3 | 389 | |||
AK220294.1 | - | 3 | 533 | |||
BC015278.1 | - | 2 | 164 | 4221290 | ||
BY724037.1 | - | 3 | 591 |