Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138652.2 | NP_619593.2 | 2 | 311 | ||
GenBank mRNA | BC109010.1 | - | 2 | 311 | ||
BC109011.1 | - | 2 | 311 | 40048684 | ||
S83328.1 | - | 1 | 95 |