Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138676.2 | NP_619617.2 | 6 | 393 | ||
GenBank mRNA | AF246218.1 | - | 6 | 399 | ||
AK047434.1 | - | 4 | 466 | |||
AK154779.1 | - | 6 | 382 | |||
AK160641.1 | - | 5 | 465 | |||
AK166328.1 | - | 5 | 338 | |||
BB847999.1 | - | 6 | 396 | |||
BC061477.1 | - | 6 | 346 | 6336438 |