Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000547.5 | NP_000538.3 | 17 | 3048 | ||
NM_001206744.1 | NP_001193673.1 | 17 | 3043 | |||
NM_001206745.1 | NP_001193674.1 | 16 | 2872 | |||
NM_175719.3 | NP_783650.1 | 16 | 2877 | |||
NM_175721.3 | NP_783652.1 | 15 | 2826 | |||
NM_175722.3 | NP_783653.1 | 15 | 2439 | |||
XM_011510380.2 | XP_011508682.1 | 15 | 2828 | |||
XM_011510381.2 | XP_011508683.1 | 14 | 2655 | |||
GenBank mRNA | AB208960.1 | - | 12 | 4310 | ||
AK309868.1 | - | 6 | 1118 | |||
BC095448.1 | - | 17 | 3041 | 30329791 | ||
J02969.1 | - | 17 | 3029 | |||
J02970.1 | - | 16 | 2858 | |||
M17755.2 | - | 17 | 3041 | |||
M55702.1 | - | 6 | 950 | |||
Y00406.1 | - | 18 | 2997 |