Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou4f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138945.2 | NP_620395.2 | 2 | 180 | ||
GenBank mRNA | AK039639.1 | - | 54 | |||
BC145941.1 | - | 2 | 180 | 40131244 |