Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290535.1 | NP_001277464.1 | 4, 5 | 577 | ||
NM_139144.4 | NP_631883.2 | 4, 5 | 774 | |||
GenBank mRNA | AF363030.1 | - | 4, 5 | 539 | ||
AK165328.1 | - | 1 | 3076 | |||
BC038435.1 | - | 1 | 833 | |||
BC048239.1 | - | 1 | 530 | |||
BC057319.1 | - | 4, 5 | 697 | 6830845 | ||
CB520299.1 | - | 4, 5 | 524 |