Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139221.2 | NP_631967.1 | 2 | 185 | ||
GenBank mRNA | AJ437646.1 | - | 2 | 164 | ||
BC099849.1 | - | 2 | 153 | 40038952 | ||
CJ044726.1 | - | 2 | 186 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK016897.1 | - | 785 | Chr. - - [-] on Build |