Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138673.2 | NP_619614.1 | 4 | 478 | ||
GenBank mRNA | AB105393.1 | - | 4 | 478 | ||
AB105394.1 | - | 4 | 547 | |||
AB105395.1 | - | 4 | 531 | |||
AB105396.1 | - | 4 | 523 | |||
AB105397.1 | - | 4 | 555 | |||
AB105398.1 | - | 4 | 523 | |||
AB105399.1 | - | 4 | 523 | |||
AF364951.2 | - | 4 | 478 | |||
BC145110.1 | - | 3 | 305 | |||
DQ786775.1 | - | 4 | 474 |