Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170848.1 | NP_001164319.1 | 5 | 805 | ||
NM_001170849.1 | NP_001164320.1 | 5 | 805 | |||
NM_138680.2 | NP_619621.2 | 5 | 805 | |||
GenBank mRNA | AF318301.1 | - | 5 | 730 | ||
AK046307.1 | - | 6 | 1003 | |||
AK134191.1 | - | 5 | 719 | |||
AK147889.1 | - | 8 | 976 | |||
AK153527.1 | - | 9 | 1062 | |||
BC020058.1 | - | 6 | 530 | 4009362 | ||
BC040793.1 | - | 4 | 297 | |||
BC054449.1 | - | 4 | 748 | |||
BC056354.1 | - | 5 | 803 | 6811460 | ||
BC056383.1 | - | 5 | 784 | 6853955 | ||
BC056970.1 | - | 5 | 803 | 6811460 |