Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004455.2 | NP_001004455.2 | 2 | 1199 | ||
NM_001161807.1 | NP_001155279.1 | 2 | 1199 | |||
NM_138686.3 | NP_619627.3 | 2 | 1199 | |||
GenBank mRNA | AF390548.1 | - | 2 | 1217 | ||
AK013490.1 | - | 2 | 360 | |||
BC139298.1 | - | 2 | 427 | 8862320 | ||
BC139302.1 | - | 2 | 427 | 8862324 | ||
BC145598.1 | - | 2 | 427 | 9054164 | ||
BC171971.1 | - | 2 | 427 | 9054257 | ||
BE996554.1 | - | 2 | 173 |