Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139001.2 | NP_620570.2 | 3 | 668 | ||
GenBank mRNA | AF352400.1 | - | 3 | 612 | ||
AK052394.1 | - | 3 | 740 |