Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgre4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139138.3 | NP_631877.2 | 7 | 619 | ||
GenBank mRNA | AF396935.1 | - | 7 | 577 | ||
AK036459.1 | - | 7 | 620 | |||
AK037483.1 | - | 7 | 564 | |||
AK155935.1 | - | 7 | 567 | |||
AY032690.1 | - | 7 | 538 | |||
BC120538.1 | - | 7 | 576 | 40129461 | ||
BC120540.1 | - | 7 | 576 | 40129463 |
Set Membership: |
G-protein coupled receptor |