Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000574.4 | NP_000565.1 | 10 | 2017 | ||
NM_001114752.2 | NP_001108224.1 | 11 | 2135 | |||
NM_001300903.1 | NP_001287832.1 | 11 | 2132 | |||
NM_001300904.1 | NP_001287833.1 | 11 | 2110 | |||
NR_125349.1 | 11 | 2102 | ||||
GenBank mRNA | AY055757.1 | - | 6 | 1525 | ||
AY055758.1 | - | 7 | 1848 | |||
M15799.1 | - | 10 | 1703 | |||
M30142.1 | - | 11 | 1906 | |||
M31516.1 | - | 10 | 1788 |