Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138673.2 | NP_619614.1 | 12, 13 | 1494 | ||
GenBank mRNA | AB105393.1 | - | 12, 13 | 1494 | ||
AB105396.1 | - | 12, 13 | 1539 | |||
AB105397.1 | - | 12, 13 | 1571 | |||
AB105398.1 | - | 12, 13 | 1539 | |||
AB105399.1 | - | 12, 13 | 1539 | |||
AF364951.2 | - | 12, 13 | 1494 | |||
BC145110.1 | - | 11, 12 | 1321 | |||
DQ786775.1 | - | 12, 13 | 1490 |