Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138676.2 | NP_619617.2 | 12 | 1137 | ||
GenBank mRNA | AF246218.1 | - | 12 | 1143 | ||
AK047434.1 | - | 10 | 1268 | |||
AK154779.1 | - | 12 | 1125 | |||
AK160641.1 | - | 11 | 1209 | |||
AK198381.1 | - | 2 | 131 | |||
AK203091.1 | - | 2 | 87 | |||
BC061477.1 | - | 12 | 1090 | 6336438 |