Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000583.3 | NP_000574.2 | 6, 7 | 1047 | ||
NM_001204306.1 | NP_001191235.1 | 7, 8 | 918 | |||
NM_001204307.1 | NP_001191236.1 | 7, 8 | 855 | |||
XM_006714177.2 | XP_006714240.1 | 6, 7 | 910 | |||
GenBank mRNA | AK223458.1 | - | 7, 8 | 918 | ||
AK290827.1 | - | 6, 7 | 757 | |||
AK298433.1 | - | 5, 6 | 615 | |||
AK309595.1 | - | 7, 8 | 855 | |||
AK315853.1 | - | 6, 7 | 910 | |||
BC057228.1 | - | 6, 7 | 765 | 4767205 | ||
EU794674.1 | - | 6, 7 | 856 | |||
X03178.1 | - | 6, 7 | 856 |