Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253768.1 | NP_001240697.1 | 15 | 2876 | ||
NM_001253769.1 | NP_001240698.1 | 14 | 2772 | |||
NM_001253770.1 | NP_001240699.1 | 14 | 2621 | |||
NM_138744.3 | NP_620083.1 | 14 | 2624 | |||
GenBank mRNA | AF532969.1 | - | 13 | 2419 | ||
AK031356.1 | - | 15 | 2600 | |||
AK043865.1 | - | 14 | 2725 | |||
AK173064.1 | - | 14 | 2611 | |||
BC021749.1 | - | 14 | 2579 | 4165430 | ||
BC031527.1 | - | 14 | 2587 | 4008624 |