Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027874.2 | NP_082150.1 | 5, 6 | 1057 | ||
NM_139059.2 | NP_620690.1 | 5, 6 | 1057 | |||
GenBank mRNA | AK004606.1 | - | 5, 6 | 817 | ||
AK051052.1 | - | 5, 6 | 1056 | |||
AK088642.1 | - | 5, 6 | 792 | |||
AK140317.1 | - | 5, 6 | 782 | |||
AK143534.1 | - | 5, 6 | 758 | |||
AK146600.1 | - | 5, 6 | 1047 | |||
AK150387.1 | - | 5, 6 | 786 | |||
AK150815.1 | - | 5, 6 | 784 | |||
AK152721.1 | - | 5, 6 | 999 | |||
AK157812.1 | - | 5, 6 | 792 | |||
AK180857.1 | - | 1, 2 | 132 | |||
AK218352.1 | - | 1, 2 | 81 | |||
BC004604.1 | - | 5, 6 | 1043 | 3585286 |