Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139140.1 | NP_631879.1 | 3, 4 | 422 | ||
GenBank mRNA | AB079132.1 | - | 3, 4 | 422 | ||
AK048720.1 | - | 3, 4 | 464 | |||
AK135838.1 | - | 3, 4 | 480 | |||
AK159335.1 | - | 3, 4 | 516 | |||
AK167788.1 | - | 2, 3 | 99 | |||
AK171266.1 | - | 2, 3 | 177 | |||
BC023432.1 | - | 1, 2 | 72 | 5029821 | ||
BC072579.1 | - | 3, 4 | 689 |