Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290535.1 | NP_001277464.1 | 21 | 2968 | ||
NM_139144.4 | NP_631883.2 | 21 | 3165 | |||
GenBank mRNA | AF363030.1 | - | 21 | 2930 | ||
AK045910.1 | - | 6 | 915 | |||
AK047095.1 | - | 2 | 252 | |||
BC038435.1 | - | 17 | 3224 | |||
BC048239.1 | - | 17 | 2920 | |||
BC057319.1 | - | 21 | 3088 | 6830845 |