Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139145.4 | NP_631884.1 | 3 | 1108 | ||
GenBank mRNA | AB066227.1 | - | 2 | 968 | ||
AK052424.1 | - | 4 | 1404 | |||
AK158212.1 | - | 4 | 1225 | |||
AK164351.1 | - | 4 | 1389 | |||
BC050090.1 | - | 3 | 1109 | 30012868 |