Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139197.2 | NP_631936.2 | 7 | 1010 | ||
GenBank mRNA | AB084789.1 | - | 6 | 759 | ||
AF292399.1 | - | 6 | 739 | |||
BC106831.2 | - | 6 | 739 | 40044790 |