Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139200.4 | NP_631939.1 | 8 | 1329 | ||
GenBank mRNA | AF192525.1 | - | 8 | 1249 | ||
AK089364.1 | - | 8 | 1270 | |||
AK152784.1 | - | 8 | 1311 | |||
AK157859.1 | - | 8 | 1330 | |||
AK169575.1 | - | 8 | 1315 | |||
AK170544.1 | - | 8 | 1330 | |||
AK170796.1 | - | 8 | 1330 | |||
AK171467.1 | - | 8 | 1330 | |||
AK172071.1 | - | 8 | 1386 | |||
BC007144.1 | - | 8 | 1318 | 3487179 |