Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139272.2 | NP_644678.2 | 1, 2 | 260 | ||
GenBank mRNA | AF348968.1 | - | 1, 2 | 128 | ||
AK046567.1 | - | 1, 2 | 218 | |||
AK145122.1 | - | 1, 2 | 138 | |||
AK149609.1 | - | 1, 2 | 186 | |||
AK151536.1 | - | 1, 2 | 51 | |||
BC059818.1 | - | 1, 2 | 205 | 5697789 | ||
BQ770019.1 | - | 1, 2 | 260 |