Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139311.2 | NP_647472.2 | 7, 8 | 921 | ||
GenBank mRNA | AK143477.1 | - | 3, 4 | 364 | ||
AY050217.1 | - | 7, 8 | 1068 | |||
BY731745.1 | - | 3, 4 | 360 |