Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 3 | 287 | ||
GenBank mRNA | AK048537.1 | - | 3 | 207 | ||
AK145292.1 | - | 3 | 319 | |||
AK161695.1 | - | 3 | 226 | |||
AK166059.1 | - | 3 | 266 | |||
AK170306.1 | - | 3 | 271 | |||
AK170669.1 | - | 3 | 288 | |||
BC026804.1 | - | 3 | 294 | 4159217 |