Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2900011O08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144518.3 | NP_653101.1 | 2, 3 | 383 | ||
GenBank mRNA | AK019307.1 | - | 2, 3 | 376 | ||
AK035868.1 | - | 2, 3 | 360 | |||
AK049997.1 | - | 2, 3 | 415 | |||
AK076136.1 | - | 2, 3 | 402 | |||
AK136859.1 | - | 2, 3 | 383 | |||
AK139470.1 | - | 2, 3 | 395 | |||
BC022741.1 | - | 2, 3 | 362 | 4511454 |