Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144522.5 | NP_653105.3 | 3 | 1202 | ||
GenBank mRNA | AF285112.1 | - | 3 | 560 | ||
AK047320.1 | - | 3 | 1203 | |||
AK047327.1 | - | 3 | 1203 | |||
BC072576.1 | - | 3 | 745 | 6401332 |