Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144527.3 | NP_653110.3 | 3 | 240 | ||
GenBank mRNA | AK010621.1 | - | 3 | 240 | ||
AK030742.1 | - | 3 | 205 | |||
AK085545.1 | - | 3 | 187 | |||
AK207753.1 | - | 3 | 116 | |||
BC008221.1 | - | 3 | 192 | |||
BC031729.1 | - | 3 | 136 | 3992639 | ||
BY250554.1 | - | 3 | 241 |