Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122640.1 | NP_001116112.1 | 3 | 283 | ||
NM_001122641.1 | NP_001116113.1 | 3 | 283 | |||
NM_001122642.1 | NP_001116114.1 | 3 | 283 | |||
NM_001122643.1 | NP_001116115.1 | 3 | 283 | |||
NM_144529.2 | NP_653112.2 | 3 | 283 | |||
GenBank mRNA | AB060553.1 | - | 3 | 281 | ||
AB060554.1 | - | 3 | 281 | |||
AK036468.1 | - | 3 | 220 | |||
AK036617.1 | - | 3 | 220 | |||
AK046411.1 | - | 3 | 272 | |||
AK147010.1 | - | 3 | 254 | |||
AK149986.1 | - | 3 | 255 | |||
AK152266.1 | - | 3 | 220 | |||
AK152301.1 | - | 3 | 247 | |||
AK161636.1 | - | 3 | 284 | |||
AK208773.1 | - | 1 | 74 | |||
BC003259.1 | - | 3 | 231 | 3500822 |