Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144532.2 | NP_653115.1 | 1 | 162 | ||
GenBank mRNA | AK171863.1 | - | 5 | 772 | ||
AY039218.1 | - | 1 | 157 | |||
BC029002.1 | - | 1 | 70 | |||
BC049263.1 | - | 1 | 162 | 5366999 | ||
BC053330.1 | - | 1 | 117 |