Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144539.5 | NP_653122.2 | 3 | 602 | ||
GenBank mRNA | AB196816.1 | - | 2 | 149 | ||
AB196817.1 | - | 2 | 149 | |||
AB196818.1 | - | 2 | 149 | |||
AB196819.1 | - | 2 | 149 | |||
AB196837.1 | - | 2 | 149 | |||
AF467909.1 | - | 2 | 149 | |||
AF467910.1 | - | 2 | 149 | |||
AF467911.1 | - | 2 | 149 | |||
AK016183.1 | - | 3 | 406 | |||
AK030135.1 | - | 3 | 463 | |||
AK030148.1 | - | 3 | 603 | |||
AK040678.1 | - | 2 | 230 | |||
AK089525.1 | - | 2 | 230 | |||
AK172665.1 | - | 2 | 196 | |||
BC011154.1 | - | 2 | 161 | 4168309 |