Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144550.4 | NP_653133.3 | 2, 3 | 197 | ||
GenBank mRNA | AK048789.1 | - | 2, 3 | 141 | ||
AY099108.1 | - | 2, 3 | 477 | |||
BC052381.1 | - | 2, 3 | 151 | 5703283 | ||
BC056960.1 | - | 2, 3 | 151 | 5703283 | ||
CX568108.1 | - | 2, 3 | 197 |