Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144552.3 | NP_653135.2 | 2 | 415 | ||
GenBank mRNA | AK158779.1 | - | 2 | 301 | ||
BC024598.1 | - | 2 | 297 | 4020199 |