Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144790.1 | NP_659039.1 | 2, 3 | 286 | ||
GenBank mRNA | AK044543.1 | - | 3, 4 | 471 | ||
AK044546.1 | - | 2, 3 | 264 | |||
BC022726.1 | - | 2, 3 | 286 | 4504539 | ||
FJ895380.1 | - | 3, 4 | 447 |