Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144794.2 | NP_659043.1 | 3 | 279 | ||
GenBank mRNA | AK049360.1 | - | 3 | 273 | ||
AK144899.1 | - | 3 | 332 | |||
AK170559.1 | - | 3 | 295 | |||
AK173027.1 | - | 3 | 272 | |||
BC014795.1 | - | 3 | 252 | 3961992 | ||
BC019442.1 | - | 3 | 279 | 4036849 |