Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metrnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144797.3 | NP_659046.1 | 2 | 651 | ||
GenBank mRNA | AK034855.1 | - | 2 | 652 | ||
AK170800.1 | - | 2 | 653 | |||
AK171200.1 | - | 2 | 397 | |||
BC019776.1 | - | 2 | 606 | 3672224 | ||
BC024445.1 | - | 2 | 666 | 4974030 | ||
BC024497.1 | - | 654 | 4982563 | |||
BC026646.1 | - | 2 | 645 | 4987595 |