Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146180.1 | NP_001139652.1 | 4, 5 | 788 | ||
NM_144800.2 | NP_659049.2 | 4, 5 | 788 | |||
GenBank mRNA | AK030533.1 | - | 2, 3 | 182 | ||
AK129139.1 | - | 4, 5 | 785 | |||
AY214918.1 | - | 4, 5 | 289 | |||
BC024131.1 | - | 4, 5 | 753 | 5101888 |