Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139308.2 | NP_647469.2 | 2 | 755 | ||
GenBank mRNA | AF244543.1 | - | 1 | 112 | ||
AK044519.1 | - | 2 | 251 | |||
AK083125.1 | - | 2 | 282 | |||
AK143699.1 | - | 2 | 756 | |||
BC017524.1 | - | 2 | 745 | 4527790 | ||
BC080747.1 | - | 2 | 726 | 4237000 |