Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 6, 7 | 700 | ||
GenBank mRNA | AK048537.1 | - | 6, 7 | 620 | ||
AK145292.1 | - | 6, 7 | 732 | |||
AK161695.1 | - | 6, 7 | 639 | |||
AK166059.1 | - | 6, 7 | 679 | |||
AK170306.1 | - | 6, 7 | 684 | |||
AK170669.1 | - | 6, 7 | 701 | |||
AK185626.1 | - | 1, 2 | 90 | |||
AK220257.1 | - | 19 | ||||
BC026804.1 | - | 6, 7 | 707 | 4159217 |