Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144514.2 | NP_653097.2 | 2, 3 | 496 | ||
GenBank mRNA | AB076722.1 | - | 3, 4 | 1173 | ||
AK082909.1 | - | 2, 3 | 473 | |||
AK133507.1 | - | 2, 3 | 472 | |||
AK147766.1 | - | 334 | ||||
BC030052.1 | - | 2, 3 | 396 | |||
BC038641.1 | - | 2, 3 | 443 | |||
BC051210.1 | - | 2, 3 | 457 | |||
BC132083.1 | - | 2, 3 | 485 | 40130360 | ||
BC132085.1 | - | 2, 3 | 486 | 40130362 | ||
D85430.1 | - | 2, 3 | 466 |