Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144520.2 | NP_653103.1 | 4, 5 | 293 | ||
GenBank mRNA | BC005759.1 | - | 4, 5 | 293 | 2655022 |