Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144525.3 | NP_653108.2 | 11, 12 | 1322 | ||
GenBank mRNA | AK034883.1 | - | 11, 12 | 1324 | ||
AK053521.1 | - | 1612 | ||||
AK143078.1 | - | 1324 | ||||
AK165285.1 | - | 10, 11 | 1485 | |||
AK166510.1 | - | 11, 12 | 1317 | |||
AK180903.1 | - | 3, 4 | 230 | |||
BC022142.1 | - | 7, 8 | 599 | 5101205 | ||
BC022602.1 | - | 5, 6 | 468 | 4161204 | ||
BC026651.1 | - | 11, 12 | 1238 | 4987771 | ||
BC027046.1 | - | 5, 6 | 616 | 5362953 | ||
BC029150.1 | - | 11, 12 | 1310 | 4954376 |