Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam64a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144526.3 | NP_653109.2 | 4 | 706 | ||
GenBank mRNA | AK049710.1 | - | 4 | 707 | ||
AK078429.1 | - | 4 | 707 | |||
AK186477.1 | - | 1 | 41 | |||
BC020115.1 | - | 4 | 692 | 3988382 | ||
BC029115.1 | - | 4 | 692 | 3983148 |